CAS RN 2166387-64-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
BI-3812, MedChemExpress
MedChemExpress BI-3812 is potent and efficacious BCL6 inhibitor, inhibiting the BTB domain of BCL6, with an IC50 of ≤3 nM; BI-3812 has antitumor activity.