CAS RN 216312-73-1
IUPAC Navn:
3,6-dibromo-9H-fluoren-9-one
Synonymer:
3,6-dibromofluoren-9-one
Molekylvægt (g/mol):
338
Molekylformel:
C13H6Br2O
InChi Key:
XXZOHYHKWDLSFS-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)C(=O)C1=C2C=C(Br)C=C1
1
–
2
af
2
Resultater
1
3,6-Dibromo-9H-fluoren-9-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,6-Dibromo-9H-fluoren-9-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.