CAS RN 214139-20-5
IUPAC Navn:
3-amino-2-methylpropanoic acid hydrate
Synonymer:
(+)-β-aminoisobutyric acid hydrate
Molekylvægt (g/mol):
121.14
Molekylformel:
C4H11NO3
InChi Key:
DJNUQSALULMICR-UHFFFAOYNA-N
SMILES:
O.CC(CN)C(O)=O
1
–
3
af
3
Resultater
1
DL-3-Aminoisobutyric Acid Hydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
DL-3-Aminoisobutyric Acid Hydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
DL-3-Aminoisobutyric Acid Hydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.