CAS RN 213745-86-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
N2-Methyl Trimethoprim (Impurity), TRC
CAS: 213745-86-9 Molekylær formel: C15 H20 N4 O3 Molekylvægt (g/mol): 304.34 Synonym: N2-Methyl-5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine,Trimethoprim Imp. A (EP) IUPAC navn: 2-N-methyl-5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine SMIL: CNc1ncc(Cc2cc(OC)c(OC)c(OC)c2)c(N)n1
2
N2-Methyl-5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.