CAS RN 212322-56-0
IUPAC Navn:
ethyl 3-{1-[3-amino-4-(methylamino)phenyl]-N-(pyridin-2-yl)formamido}propanoate
Synonymer:
ethyl 3-{1-[3-amino-4-(methylamino)phenyl]-N-(pyridin-2-yl)formamido}propanoate
Molekylvægt (g/mol):
342.4
Molekylformel:
C18H22N4O3
InChi Key:
PCPATNZTKBOKOY-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCN(C(=O)C1=CC(N)=C(NC)C=C1)C1=CC=CC=N1
1
–
4
af
4
Resultater
1
N-[3-Amino-4-(methylamino)benzoyl]-N-2-pyridinyl-Beta-alanine Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[3-Amino-4-(methylamino)benzoyl]-N-2-pyridinyl-Beta-alanine Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-[3-Amino-4-(methylamino)benzoyl]-N-2-pyridinyl-b-alanine Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Ethyl 3-[[3-Amino-4-(methylamino)benzoyl]-(2-pyridyl)amino]propanoate (N-[3-Amino-4-(methylamino)benzoyl]-N-2-pyridinyl-beta-alanine Ethyl Ester), Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.