CAS RN 2122-70-5
IUPAC Navn:
ethyl 2-(naphthalen-1-yl)acetate
Synonymer:
ethyl 2-(naphthalen-1-yl)acetate
Molekylvægt (g/mol):
214.26
Molekylformel:
C14H14O2
InChi Key:
XIDPSKQLXKCVQN-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC1=C2C=CC=CC2=CC=C1
1
–
1
af
1
Resultater
1
Ethyl (1-Naphthyl)acetate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.