CAS RN 21161-11-5
IUPAC Navn:
2-nitrobenzene-1,3-dicarboxylic acid
Synonymer:
2-nitrobenzene-1,3-dicarboxylic acid
Molekylvægt (g/mol):
211.13
Molekylformel:
C8H5NO6
InChi Key:
CAHWDGJDQYAFHM-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC(C(O)=O)=C1[N+]([O-])=O
1
–
2
af
2
Resultater
1
2-Nitroisophthalic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Nitroisophthalic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.