Få mere at vide
2-Nitroisophthalic Acid, TRC
Artikelnummer. 30475138
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475138 1 g
30467148 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30475138 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5152751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Nitroisophthalic Acid
CAS 21161-11-5
Molekylær formel C8H5NO6
Mængde 1 g
Synonym -Nitro-1,3-benzenedicarboxylic Acid, 2-Nitroisophthalic Acid, 3-Carboxy-2-nitrobenzoic Acid
InChI formel InChI=1S/C8H5NO6/c10-7(11)4-2-1-3-5(8(12)13)6(4)9(14)15/h1-3H,(H,10,11)(H,12,13)
SMIL OC(=O)c1cccc(C(=O)O)c1[N+](=O)[O-]
IUPAC navn 2-nitrobenzene-1,3-dicarboxylic acid
Molekylvægt (g/mol) 211.13
Formel vægt 211.0117