CAS RN 211513-37-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Dalcetrapib, MedChemExpress
MedChemExpress Dalcetrapib (JTT-705; RO-4607381) is a rhCETP inhibitor with IC50 of 0.2 μM that increases the plasma HDL cholesterol.