missing translation for 'onlineSavingsMsg'
Få mere at vide

Dalcetrapib, MedChemExpress

Product Code. 16476606
Encompass_Preferred
Change view
Click to view available options
Mængde:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
16476606 100 mg 100mg
16432397 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16476606 Supplier Medchem Express Supplier No. HY14950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dalcetrapib (JTT-705; RO-4607381) is a rhCETP inhibitor with IC50 of 0.2 μM that increases the plasma HDL cholesterol.

Dalcetrapib (JTT-705

Specifications

Kemisk navn eller materiale Dalcetrapib
Fysisk form Solid
CAS 211513-37-0
Farve White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C23H35NO2S
Til brug med (applikation) COVID-19-immunoregulation
Synonym JTT-705 RO4607381
Opløselighedsinformation DMSO : ≥ 50 mg/mL (128.34 mM)
SMIL CC(C)C(SC1=C(NC(C2(CC(CC)CC)CCCCC2)=O)C=CC=C1)=O
Molekylvægt (g/mol) 389.59
Formel vægt 389.59
Procent renhed 99.19%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.