CAS RN 210357-36-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
4-Methylumbelliferyl 2-Acetamido-2-deoxy-3-O-(Beta-D-galactopyranosyl)-Alpha-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Methylumbelliferyl 2-Acetamido-2-deoxy-3-O-(Beta-D-galactopyranosyl)-Alpha-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Methylumbelliferyl 2-Acetamido-2-deoxy-3-O-(b-D-galactopyranosyl)-a-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.