CAS RN 2102609-59-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(1RS)-1-[3-(Dimethylamino)propyl]-1-hydroxy-1,3-dihydro-2-benzofuran-5-carbonitrile, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.