CAS RN 2098836-50-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
AT6, MedChemExpress
MedChemExpress AT6 is a PROTAC AT1 analogue, which is a PROTAC connected by ligands for von Hippel-Lindau and BRD4 with highly selectivity to bromodomain (Brd4).