CAS RN 2086274-75-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
a,a'-[(Methylimino)di-3,1-propanediyl]bis[3,4-dimethoxy-a-(1-methylethyl)-benzeneacetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
a,a'-[(Methylimino)di-3,1-propanediyl]bis[3,4-dimethoxy-a-(1-methylethyl)-benzeneacetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
a,a'-[(Methylimino)di-3,1-propanediyl]bis[3,4-dimethoxy-a-(1-methylethyl)-benzeneacetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.