CAS RN 2083627-02-3
IUPAC Navn:
N-[(furan-2-yl)methyl]-8-(4-methanesulfonylphenyl)-[1,2,4]triazolo[4,3-c]pyrimidin-5-amine
Synonymer:
N-(furan-2-ylmethyl)-8-(4-methanesulfonylphenyl)-[1,2,4]triazolo[4,3-c]pyrimidin-5-amine
Molekylvægt (g/mol):
369.4
Molekylformel:
C17H15N5O3S
InChi Key:
DYIRSNMPIZZNBK-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)C1=CC=C(C=C1)C1=CN=C(NCC2=CC=CO2)N2C=NN=C12
1
–
3
af
3
Resultater
2
Tocris Bioscience™ EED 226
Potent and selective EED inhibitor
3
EED226, MedChemExpress
MedChemExpress EED226 is a polycomb repressive complex 2 (PRC2) inhibitor, which binds to the K27me3-pocket on embryonic ectoderm development (EED) and shows strong antitumor activity in xenograft mice model. EED226 is a potent, selective, and orally bioavailable EED inhibitor. EED226 inhibits PRC2 with an IC50 of 23.4 nM when the H3K27me0 peptide is used as a substrate in the in vitro enzymatic assays.