CAS RN 207122-16-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
BDE 183, TRC
CAS: 207122-16-5 Molekylær formel: C12 H3 Br7 O Molekylvægt (g/mol): 722.48 Synonym: Benzene, 1,2,3,5-tetrabromo-4-(2,4,5-tribromophenoxy)-,1,2,3,5-Tetrabromo-4-(2,4,5-tribromophenoxy)benzene,2,2',3,4,4',5',6-Heptabromodiphenyl ether,BDE 183,PBDE 183,PBDE No. 183 IUPAC navn: 1,2,3,5-tetrabromo-4-(2,4,5-tribromophenoxy)benzene SMIL: Brc1cc(Br)c(Oc2c(Br)cc(Br)c(Br)c2Br)cc1Br
2
PBDE 183 50 μg/mL in Nonane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality