CAS RN 2070014-88-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
SRI-011381 hydrochloride, MedChemExpress
MedChemExpress SRI-011381 hydrochloride is an orally active TGF-β signaling agonist, exhibits neuroprotective effects.