CAS RN 2070014-78-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Mardepodect hydrochloride, MedChemExpress
MedChemExpress Mardepodect hydrochloride (PF-2545920 hydrochloride) is a potent, orally active and selective PDE10A inhibitor with an IC50 of 0.37 nM, with >1000-fold selectivity over other PDEs. Mardepodect hydrochloride can cross the blood-brain barrier.