CAS RN 2070009-31-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(R,S)-Ivosidenib, MedChemExpress
MedChemExpress (R,S)-Ivosidenib ((R,S)-AG-120) is the less active enantiomer of Ivosidenib (AG-120).