CAS RN 2055397-28-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ WM 1119
High affinity KAT6A (MOZ) competitive inhibitor
2
WM-1119, MedChemExpress
MedChemExpress WM-1119 is a highly potent and selective KAT6A inhibitor, with an IC50 of 0.25 μM for KAT6A in lymphoma cells, the binding KD values of WM-1119 with KAT6A, KAT5 and KAT7 are 2 nM, 2.2 μM, 0.5 μM , respectively.