CAS RN 20485-43-2
IUPAC Navn:
1-methyl-1H-imidazole-2-carboxylate
Synonymer:
1-methylimidazole-2-carboxylate
Molekylvægt (g/mol):
125.11
Molekylformel:
C5H5N2O2
InChi Key:
WLDPWZQYAVZTTP-UHFFFAOYSA-M
SMILES:
CN1C=CN=C1C([O-])=O
1
–
2
af
2
Resultater
1
1-Methylimidazole-2-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Methylimidazole-2-carboxylic acid hydrate, 90+%, Thermo Scientific™
CAS: 20485-43-2 Molekylær formel: C5H6N2O2 Molekylvægt (g/mol): 126.12 MDL nummer: MFCD01863421 InChI nøgle: WLDPWZQYAVZTTP-UHFFFAOYSA-N Synonym: 1-methyl-1h-imidazole-2-carboxylic acid,1h-imidazole-2-carboxylic acid, 1-methyl,1-methyl-1h-2-imidazolecarboxylic acid,pubchem8048,acmc-209fas,meimd 2-oh,ksc205s8r,1-methylimidazole-2-carboxylicacid,n-methyl-imidazole-2-carboxylic acid,1-methyl-imidazole-2-carboxylic acid PubChem CID: 557678 ChEBI: CHEBI:74743 IUPAC navn: 1-methylimidazole-2-carboxylic acid SMIL: CN1C=CN=C1C(O)=O