CAS RN 204255-02-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(1R,5R,6R)-1-(1-Ethylpropoxy)-7-azabicyclo[4.1.0]hept-3-ene-3-carboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.