CAS RN 203923-89-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Karenitecin, MedChemExpress
MedChemExpress Karenitecin (Cositecan) is a topoisomerase I inhibitor, with potent anti-cancer activity.