CAS RN 2036044-77-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Porcn-IN-1, MedChemExpress
MedChemExpress Porcn-IN-1 is potent porcupine inhibitor with an IC50 of 0.5±0.2 nM.