CAS RN 2032-59-9
IUPAC Navn:
4-(dimethylamino)-3-methylphenyl N-methylcarbamate
Synonymer:
aminocarb matacil
Molekylvægt (g/mol):
208.26
Molekylformel:
C11H16N2O2
InChi Key:
IMIDOCRTMDIQIJ-UHFFFAOYSA-N
SMILES:
CNC(=O)OC1=CC=C(N(C)C)C(C)=C1