CAS RN 2025363-77-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(3a,5b,6a)-6-(Ethyl-1,1,2,2,2-d5)-3-hydroxy-7-oxocholan-24-oic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3Alpha,5Beta,6Alpha)-6-(Ethyl-1,1,2,2,2-d5)-3-hydroxy-7-oxocholan-24-oic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3Alpha,5Beta,6Alpha)-6-(Ethyl-1,1,2,2,2-d5)-3-hydroxy-7-oxocholan-24-oic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.