Få mere at vide
(3a,5b,6a)-6-(Ethyl-1,1,2,2,2-d5)-3-hydroxy-7-oxocholan-24-oic Acid, TRC
Artikelnummer. 30374159
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30374159 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374159 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9458222.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3alpha,5beta,6alpha)-6-(Ethyl-1,1,2,2,2-d5)-3-hydroxy-7-oxocholan-24-oic Acid
CAS 2025363-77-1
Molekylær formel C26H37D5O4
Mængde 2.5 mg
InChI formel InChI=1S/C26H42O4/c1-5-17-21-14-16(27)10-12-26(21,4)20-11-13-25(3)18(15(2)6-9-22(28)29)7-8-19(25)23(20)24(17)30/h15-21,23,27H,5-14H2,1-4H3,(H,28,29)/t15-,16-,17-,18-,19+,20+,21+,23+,25-,26-/m1/s1/i1D3,5D2
SMIL [2H]C([2H])([2H])C([2H])([2H])[C@@H]1[C@@H]2C[C@H](O)CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3C1=O)[C@H](C)CCC(=O)O
IUPAC navn (4R)-4-[(3R,5S,6R,8S,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-7-oxo-6-(1,1,2,2,2-pentadeuterioethyl)-1,2,3,4,5,6,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvægt (g/mol) 423.64
Formel vægt 423.3397