CAS RN 202325-50-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
o-Cresol-d7, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
o-Cresol-d7, TRC
CAS: 202325-50-6 Molekylær formel: C7 2H7 H O Molekylvægt (g/mol): 115.18 Synonym: Phen-2,3,4,5-d4-ol, 6-(methyl-d3)- (9CI),2,3,4,5-Tetradeuterio-6-(trideuteriomethyl)phenol,2-(Methyl-d3)phenol-3,4,5,6-d4,o-Cresol-d7,2-Methylphenol-D7 IUPAC navn: 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)phenol SMIL: [2H]c1c([2H])c([2H])c(c(O)c1[2H])C([2H])([2H])[2H]