CAS RN 20194-18-7
IUPAC Navn:
sodium phenylmethanolate
Synonymer:
sodium phenylmethanolate
Molekylvægt (g/mol):
130.12
Molekylformel:
C7H7NaO
InChi Key:
LFQULJPVXNYWAG-UHFFFAOYSA-N
SMILES:
[Na+].[O-]CC1=CC=CC=C1
1
–
1
af
1
Resultater
1
Natriumbenzyloxid, 1 M opløsning i benzylalkohol, AcroSeal™ , Thermo Scientific Chemicals
CAS: 20194-18-7 | C7H7NaO | 130.12 g/mol