CAS RN 19982-07-1
IUPAC Navn:
N-(3,5-dimethyladamantan-1-yl)acetamide
Synonymer:
N-(3,5-dimethyladamantan-1-yl)acetamide
Molekylvægt (g/mol):
221.34
Molekylformel:
C14H23NO
InChi Key:
WVIRSYCDAYUOMJ-UHFFFAOYNA-N
SMILES:
CC(=O)NC12CC3CC(C)(CC(C)(C3)C1)C2
1
–
2
af
2
Resultater
1
N-Acetylmemantine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-Acetyl Memantine, TRC
CAS: 19982-07-1 Molekylær formel: C14 H23 N O Molekylvægt (g/mol): 221.34 Synonym: Acetamide, N-(3,5-dimethyltricyclo[3.3.1.13,7]dec-1-yl)-,Acetamide, N-(3,5-dimethyl-1-adamantyl)- (7CI,8CI),N-(3,5-Dimethyltricyclo[3.3.1.13,7]dec-1-yl)acetamide,1-Acetamido-3,5-dimethyladamantane,1-Acetylamino-3,5-dimethyladamantane,N-Acetylmemantine IUPAC navn: N-(3,5-dimethyl-1-adamantyl)acetamide SMIL: CC(=O)NC12CC3CC(C)(CC(C)(C3)C1)C2