CAS RN 19757-97-2
IUPAC Navn:
methyl 2-(hydroxymethoxy)acetate
Synonymer:
methyl 2-(hydroxymethoxy)acetate
Molekylvægt (g/mol):
120.1
Molekylformel:
C4H8O4
InChi Key:
RNVVYVPDHMKDTL-UHFFFAOYSA-N
SMILES:
COC(=O)COCO
1
–
1
af
1
Resultater
1
Methyl 2-Hydroxy-2-methoxyacetate, Technical Grade, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.