CAS RN 194798-83-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Fosfluconazole, MedChemExpress
MedChemExpress Fosfluconazole is a prodrug of Fluconazole that is widely used as an antifungal agent.