CAS RN 190791-29-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Lasofoxifene tartrate, MedChemExpress
MedChemExpress Lasofoxifene (CP-336156) tartrate is an orally active and selective estrogen receptor modulator (SERM). Lasofoxifene tartrate exhibits an anti-osteoporotic function and also inhibits primary tumor growth and metastases. Lasofoxifene tartrate can be used for the research of breast cancer and postmenopausal osteoporosis.