CAS RN 1904651-39-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
7-(Difluoromethyl)-6-(1-methylpyrazol-4-yl)-1,2,3,4-tetrahydroquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-(Difluoromethyl)-6-(1-methylpyrazol-4-yl)-1,2,3,4-tetrahydroquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-(Difluoromethyl)-6-(1-methylpyrazol-4-yl)-1,2,3,4-tetrahydroquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.