CAS RN 189-64-0
IUPAC Navn:
hexacyclo[10.10.2.0²,⁷.0⁹,²³.0¹³,¹⁸.0²⁰,²⁴]tetracosa-1(23),2,4,6,8,10,12(24),13,15,17,19,21-dodecaene
Synonymer:
dibenzo[a,h]pyrene
Molekylvægt (g/mol):
302.38
Molekylformel:
C24H14
InChi Key:
RXUSYFJGDZFVND-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C=C1C=CC3=C4C(C=CC2=C14)=CC1=CC=CC=C31
1
–
4
af
4
Resultater
1
Dibenzo[b,def]chrysene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dibenzo[b,def]chrysene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.