CAS RN 1887015-55-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
5-Fluoro-1-methyl-6-oxo-1,6-dihydropyridine-2-carbonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Fluoro-1-methyl-6-oxo-1,6-dihydropyridine-2-carbonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Fluoro-1-methyl-6-oxo-1,6-dihydropyridine-2-carbonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.