CAS RN 186204-32-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Lurasidone Metabolite 14283 hydrochloride, MedChemExpress
MedChemExpress Lurasidone Metabolite 14283 hydrochloride is a major active metabolite of Lurasidone. Lurasidone is a FDA approved drug for the treatment of schizophrenia.