CAS RN 18395-90-9
IUPAC Navn:
di-tert-butyldichlorosilane
Synonymer:
di-tert-butyldichlorosilane
Molekylvægt (g/mol):
213.22
Molekylformel:
C8H18Cl2Si
InChi Key:
PDYPRPVKBUOHDH-UHFFFAOYSA-N
SMILES:
CC(C)(C)[Si](Cl)(Cl)C(C)(C)C
1
–
3
af
3
Resultater
1
Di-tert-butyldichlorosilane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Di-tert-butyldichlorosilane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Di-tert-butyldichlorosilane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.