CAS RN 183506-73-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
1,2,3,6-Tetra-O-acetyl-4-deoxy-4-fluoro-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2,3,6-Tetra-O-acetyl-4-deoxy-4-fluoro-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2,3,6-Tetra-O-acetyl-4-deoxy-4-fluoro-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.