CAS RN 1823864-72-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2-(tert-Butyl) 3a,6a-Dimethyl dihydropyrrolo[3,4-c]pyrrole-2,3a,6a(1H,3H,4H)-tricarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(tert-Butyl) 3a,6a-Dimethyl dihydropyrrolo[3,4-c]pyrrole-2,3a,6a(1H,3H,4H)-tricarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(tert-Butyl) 3a,6a-Dimethyl dihydropyrrolo[3,4-c]pyrrole-2,3a,6a(1H,3H,4H)-tricarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.