missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(tert-Butyl) 3a,6a-Dimethyl dihydropyrrolo[3,4-c]pyrrole-2,3a,6a(1H,3H,4H)-tricarboxylate, TRC
Specifications
Specifications
| Kemisk navn eller materiale | 2-(tert-Butyl) 3a,6a-Dimethyl dihydropyrrolo[3,4-c]pyrrole-2,3a,6a(1H,3H,4H)-tricarboxylate |
| CAS | 1823864-72-7 |
| Molekylær formel | C15 H24 N2 O6 |
| Mængde | 100 mg |
| InChI formel | InChI=1S/C15H24N2O6/c1-13(2,3)23-12(20)17-8-14(10(18)21-4)6-16-7-15(14,9-17)11(19)22-5/h16H,6-9H2,1-5H3 |
| SMIL | COC(=O)C12CNCC1(CN(C2)C(=O)OC(C)(C)C)C(=O)OC |
| IUPAC navn | 5-O-tert-butyl 3a-O,6a-O-dimethyl 2,3,4,6-tetrahydro-1H-pyrrolo[3,4-c]pyrrole-3a,5,6a-tricarboxylate |
| Molekylvægt (g/mol) | 328.361 |
| Formel vægt | 328.163 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?