CAS RN 1821143-79-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
DMU2105, MedChemExpress
MedChemExpress DMU2105 is a potent and specific CYP1B1 inhibitor, with IC50s of 10 nM and 742 nM for CYP1B1 and CYP1A1, respectively.