CAS RN 1818389-84-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
SPHINX31, MedChemExpress
MedChemExpress SPHINX31 is a potent and selective inhibitor of serine/arginine-rich protein kinase 1 (SRPK1), with an IC50 of 5.9 nM. SPHINX31 inhibits phosphorylation of serine/arginine-rich splicing factor 1 (SRSF1). SPHINX31 is a potential topical therapeutic for neovascular eye disease.