CAS RN 1803592-71-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
4-(4-Amino-3-methoxyphenyl)-1lambda6-thiomorpholine-1,1-dione Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(4-Amino-3-methoxyphenyl)-1lambda6-thiomorpholine-1,1-dione Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(4-Amino-3-methoxyphenyl)-1lambda6-thiomorpholine-1,1-dione Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.