CAS RN 1798894-89-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Benzyl N-[(2’-(Trityltetrazol-5-yl-1,1’-biphenyl-4-yl]-methyl-2-amino-3-methylbutanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzyl (2S)-3-Methyl-2-[[[2'-[2-(triphenylmethyl)tetrazol-5-yl]biphenyl-4-yl]methyl]amino]butanoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Benzyl N-[(2’-(Trityltetrazol-5-yl-1,1’-biphenyl-4-yl]-methyl-2-amino-3-methylbutanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.