CAS RN 1798014-38-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
N-[N’-Hydroyethyl-N’-[3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-2-ethylamine]-N-methyl-formamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[N’-Hydroyethyl-N’-[3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-2-ethylamine]-N-methyl-formamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.