CAS RN 1797432-62-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tyk2-IN-5, MedChemExpress
MedChemExpress Tyk2-IN-5 (compound 6) is a highly potent, selective and orally active Tyk2 inhibitor and targets the JH2 domain, with a Ki of 0.086 nM for Tyk2 JH2 and an IC50 of 25 nM for IFNα. Highly effective in inhibiting IFNγ production in a rat pharmacodynamics model and fully efficacious in a rat adjuvant arthritis model.