CAS RN 1797134-99-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
6-Chloro-4-[3-cyclopropyl-1-hydroxy-1-(trifluoromethyl)-2-propyn-1-yl]-2(3H)-benzoxazolone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Chloro-4-[3-cyclopropyl-1-hydroxy-1-(trifluoromethyl)-2-propyn-1-yl]-2(3H)-benzoxazolone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-Chloro-4-[3-cyclopropyl-1-hydroxy-1-(trifluoromethyl)-2-propyn-1-yl]-2(3H)-benzoxazolone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.