CAS RN 1797132-23-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
1,7-Di(2-methoxyphenoxy)-2,6-dihydroxy-4-oxaheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,1'-Oxybis[3-(2-methoxyphenoxy)propan-2-ol] (Bisether), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
1,7-Di(2-methoxyphenoxy)-2,6-dihydroxy-4-oxaheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.