Få mere at vide
1,1'-Oxybis[3-(2-methoxyphenoxy)propan-2-ol] (Bisether), Mikromol™
Artikelnummer. 30052521
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052521 25 mg
17950926 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052521 Leverandør Mikromol Leverandørnr. MM0354.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1'-Oxybis[3-(2-methoxyphenoxy)propan-2-ol] (Bisether)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1'-Oxybis[3-(2-methoxyphenoxy)propan-2-ol]
CAS 1797132-23-0
Analyt- eller komponentnavne 1,1'-Oxybis[3-(2-methoxyphenoxy)propan-2-ol]
Forsendelsestilstand Room Temperature
Molekylær formel C20 H26 O7
Certificeringer/compliance ISO 17025
Synonym Guaifenesin Imp. C (EP), 1,1'-Oxybis[3-(2-methoxyphenoxy)propan-2-ol], Bisether of Guaifenesin, Guaifenesin Impurity C
InChI formel InChI=1S/C12H18N2O3S.ClH/c1-4-5-13-6-9(15)14-10-8(2)7-18-11(10)12(16)17-3;/h7,13H,4-6H2,1-3H3,(H,14,15);1H
SMIL COc1ccccc1OCC(O)COCC(O)COc2ccccc2OC
IUPAC navn 1-[2-hydroxy-3-(2-methoxyphenoxy)propoxy]-3-(2-methoxyphenoxy)propan-2-ol
Molekylvægt (g/mol) 378.42
Mængde 25 mg
Formel vægt 378.1679 g/mol